Assay Details
| Gene Symbol: CBL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334162.1 | NP_001319319.1 | 5 - 6 | 469 | 67 | |
| NM_001334163.1 | NP_001322653.1 | 5 - 6 | 445 | 67 | ||
| NM_001334164.1 | NP_001322654.1 | 5 - 6 | 464 | 67 | ||
| NM_105123.2 | NP_176629.1 | 4 - 5 | 344 | 67 | ||
| GenBank mRNA | AF290433.1 | - | 4 - 5 | 344 | 67 | |
| AF411957.1 | - | 4 - 5 | 344 | 67 | ||
| BT026348.1 | - | 4 - 5 | 344 | 67 | ||
More Information
Set Membership: |
|

CBL8
Genomic Map