Assay Details
| Gene Symbol: RPN12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105127.4 | NP_176633.1 | 1 - 2 | 290 | 96 | |
| GenBank mRNA | AF410265.1 | - | 1 - 2 | 146 | 96 | |
| AY039857.1 | - | 1 - 2 | 109 | 96 | ||
| AY143888.1 | - | 1 - 2 | 100 | 96 | ||
| AY230846.1 | - | 1 - 2 | 123 | 96 | ||
More Information
Set Membership: |
|

RPN12a
Genomic Map