Assay Details
Gene Symbol: RPN12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105127.4 | NP_176633.1 | 3 - 4 | 479 | 128 | |
GenBank mRNA | AF410265.1 | - | 3 - 4 | 335 | 128 | |
AY039857.1 | - | 3 - 4 | 298 | 128 | ||
AY143888.1 | - | 3 - 4 | 289 | 128 | ||
AY230846.1 | - | 3 - 4 | 312 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
