Assay Details
Gene Symbol: SS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110984.3 | NP_186767.1 | 1 - 2 | 714 | 82 | |
GenBank mRNA | AY054467.1 | - | 1 - 2 | 382 | 82 | |
BT002555.1 | - | 1 - 2 | 281 | 82 | ||
KJ138848.1 | - | 1 - 2 | 281 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
