Assay Details
| Gene Symbol: ROPGEF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117395.3 | NP_193060.1 | 6 - 7 | 1361 | 68 | |
| GenBank mRNA | DQ446830.1 | - | 6 - 7 | 1158 | 68 | |
More Information
Set Membership: |
|

ROPGEF9
Genomic Map