Assay Details
Gene Symbol: SBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203794.2 | NP_001190723.1 | 2 - 3 | 197 | 65 | |
NM_117478.6 | NP_193139.1 | 1 - 2 | 330 | 65 | ||
GenBank mRNA | AY062634.1 | - | 1 - 2 | 203 | 65 | |
AY128727.1 | - | 1 - 2 | 155 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
