Assay Details
Gene Symbol: SBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203794.2 | NP_001190723.1 | 7 - 8 | 1232 | 95 | |
NM_117478.6 | NP_193139.1 | 6 - 7 | 1365 | 95 | ||
GenBank mRNA | AY062634.1 | - | 6 - 7 | 1238 | 95 | |
AY128727.1 | - | 6 - 7 | 1190 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
