Assay Details
Gene Symbol: PDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340907.1 | NP_001319934.1 | 12 - 13 | 1591 | 61 | |
NM_001340908.1 | NP_001329033.1 | 11 - 12 | 1439 | 61 | ||
NM_117498.4 | NP_193157.1 | 12 - 13 | 1633 | 61 | ||
GenBank mRNA | AF360196.1 | - | 12 - 13 | 1411 | 61 | |
AY040007.1 | - | 11 - 12 | 1280 | 61 | ||
L16237.1 | - | 12 - 13 | 1407 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
