Assay Details
Gene Symbol: PDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340907.1 | NP_001319934.1 | 4 - 5 | 704 | 80 | |
NM_001340908.1 | NP_001329033.1 | 3 - 4 | 552 | 80 | ||
NM_117498.4 | NP_193157.1 | 4 - 5 | 746 | 80 | ||
GenBank mRNA | AF360196.1 | - | 4 - 5 | 524 | 80 | |
AY040007.1 | - | 3 - 4 | 393 | 80 | ||
L16237.1 | - | 4 - 5 | 520 | 80 |
Gene Symbol: ACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK230311.1 | - | 72 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
