Assay Details
| Gene Symbol: ACD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117519.4 | NP_567430.1 | 2 - 3 | 457 | 68 | |
| GenBank mRNA | AK176809.1 | - | 2 - 3 | 170 | 68 | |
| AY094416.1 | - | 2 - 3 | 172 | 68 | ||
| AY344843.1 | - | 2 - 3 | 171 | 68 | ||
| BT002281.1 | - | 1 - 2 | 77 | 68 | ||
More Information
Set Membership: |
|

ACD6
Genomic Map