Assay Details
| Gene Symbol: AT4G14815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117567.4 | NP_567445.1 | 1 - 2 | 390 | 142 | |
| GenBank mRNA | AB246333.1 | - | 1 - 2 | 311 | 142 | |
| DQ446836.1 | - | 1 - 2 | 311 | 142 | ||
More Information
Set Membership: |
|

AT4G14815
Genomic Map