Assay Details
Gene Symbol: OASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203803.1 | NP_001190732.1 | 2 - 3 | 328 | 82 | |
NM_001203804.1 | NP_001190733.1 | 2 - 3 | 331 | 82 | ||
NM_001340975.1 | NP_001328013.1 | 2 - 3 | 510 | 82 | ||
NM_117574.4 | NP_193224.1 | 2 - 3 | 315 | 82 | ||
NM_179055.3 | NP_849386.1 | 2 - 3 | 185 | 82 | ||
GenBank mRNA | BT025878.1 | - | 1 - 2 | 37 | 82 | |
X80376.2 | - | 98 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
