Assay Details
| Gene Symbol: OASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203803.1 | NP_001190732.1 | 6 - 7 | 888 | 88 | |
| NM_001203804.1 | NP_001190733.1 | 6 - 7 | 891 | 88 | ||
| NM_001340975.1 | NP_001328013.1 | 6 - 7 | 1070 | 88 | ||
| NM_117574.4 | NP_193224.1 | 6 - 7 | 875 | 88 | ||
| NM_179055.3 | NP_849386.1 | 6 - 7 | 745 | 88 | ||
| GenBank mRNA | BT025878.1 | - | 5 - 6 | 597 | 88 | |
| X80376.2 | - | 658 | 88 | |||
More Information
Set Membership: |
|

OASA1
Genomic Map