Assay Details
| Gene Symbol: OASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203803.1 | NP_001190732.1 | 9 - 10 | 1084 | 66 | |
| NM_001203804.1 | NP_001190733.1 | 9 - 10 | 1087 | 66 | ||
| NM_001340975.1 | NP_001328013.1 | 9 - 10 | 1266 | 66 | ||
| NM_117574.4 | NP_193224.1 | 9 - 10 | 1071 | 66 | ||
| NM_179055.3 | NP_849386.1 | 9 - 10 | 941 | 66 | ||
| GenBank mRNA | BT025878.1 | - | 8 - 9 | 793 | 66 | |
| X80376.2 | - | 854 | 66 | |||
More Information
Set Membership: |
|

OASA1
Genomic Map