Assay Details
| Gene Symbol: FAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340992.1 | NP_001329445.1 | 6 - 7 | 2781 | 65 | |
| NM_001340993.1 | NP_001329446.1 | 5 - 6 | 3122 | 65 | ||
| NM_001340994.1 | NP_001329443.1 | 6 - 7 | 2916 | 65 | ||
| NM_001340995.1 | NP_001329444.1 | 6 - 7 | 2879 | 65 | ||
| NM_117596.6 | NP_567455.4 | 6 - 7 | 2710 | 65 | ||
| GenBank mRNA | AF159587.1 | - | 5 - 6 | 2398 | 65 | |
More Information
Set Membership: |
|

FAR1
Genomic Map