Assay Details
| Gene Symbol: BAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117609.3 | NP_567460.1 | 1 - 2 | 235 | 98 | |
| NM_179058.1 | NP_849389.1 | 1 - 2 | 179 | 98 | ||
| GenBank mRNA | AF424573.1 | - | 1 - 2 | 174 | 98 | |
| AK226353.1 | - | 1 - 2 | 177 | 98 | ||
| AK316869.1 | - | 1 - 2 | 175 | 98 | ||
| AY142024.1 | - | 1 - 2 | 66 | 98 | ||
| M73467.1 | - | 155 | 98 | |||
More Information
Set Membership: |
|

BAM5
Genomic Map