Assay Details
| Gene Symbol: BAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340998.1 | NP_001329368.1 | 2 - 3 | 854 | 104 | |
| NM_001340999.1 | NP_001329367.1 | 2 - 3 | 1027 | 104 | ||
| NM_117609.3 | NP_567460.1 | 2 - 3 | 629 | 104 | ||
| NM_179058.1 | NP_849389.1 | 2 - 3 | 573 | 104 | ||
| GenBank mRNA | AF424573.1 | - | 2 - 3 | 568 | 104 | |
| AK226353.1 | - | 2 - 3 | 571 | 104 | ||
| AK316869.1 | - | 2 - 3 | 569 | 104 | ||
| AY142024.1 | - | 2 - 3 | 460 | 104 | ||
| M73467.1 | - | 549 | 104 | |||
More Information
Set Membership: |
|

BAM5
Genomic Map