Assay Details
| Gene Symbol: UGT84A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117638.3 | NP_193283.2 | 1 | 1023 | 121 | |
| GenBank mRNA | BT015796.1 | - | 1 | 979 | 121 | |
| KJ138754.1 | - | 1 | 979 | 121 | ||
More Information
Set Membership: |
|

UGT84A1
Genomic Map