Assay Details
| Gene Symbol: UGT84A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117639.3 | NP_193284.1 | 1 | 1817 | 137 | |
| GenBank mRNA | AY057646.1 | - | 1 | 1482 | 137 | |
| AY074339.1 | - | 1 | 1467 | 137 | ||
More Information
Set Membership: |
|

UGT84A3
Genomic Map