Assay Details
| Gene Symbol: UGT84A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117640.4 | NP_193285.1 | 1 | 1089 | 61 | |
| GenBank mRNA | AK229801.1 | - | 1 | 983 | 61 | |
| BT012573.1 | - | 1 | 933 | 61 | ||
| KJ138752.1 | - | 1 | 933 | 61 | ||
More Information
Set Membership: |
|

UGT84A4
Genomic Map