Assay Details
| Gene Symbol: PRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117682.3 | NP_193325.1 | 10 - 11 | 961 | 106 | |
| GenBank mRNA | BT029023.1 | - | 10 - 11 | 852 | 106 | |
| X82825.1 | - | 874 | 106 | |||
More Information
Set Membership: |
|

PRL1
Genomic Map