Assay Details
| Gene Symbol: PRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117682.3 | NP_193325.1 | 1 - 2 | 244 | 76 | |
| GenBank mRNA | BT029023.1 | - | 1 - 2 | 135 | 76 | |
| X82825.1 | - | 157 | 76 | |||
More Information
Set Membership: |
|

PRL1
Genomic Map