Assay Details
| Gene Symbol: PRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117682.3 | NP_193325.1 | 4 - 5 | 459 | 83 | |
| GenBank mRNA | BT029023.1 | - | 4 - 5 | 350 | 83 | |
| X82825.1 | - | 372 | 83 | |||
More Information
Set Membership: |
|

PRL1
Genomic Map