Assay Details
Gene Symbol: IMPA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160767.2 | NP_001154239.1 | 2 - 3 | 691 | 79 | |
NM_117708.6 | NP_567485.4 | 2 - 3 | 691 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
