Assay Details
| Gene Symbol: AT4G16360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117731.7 | NP_193369.3 | 1 - 2 | 644 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G16360| Gene Symbol: AT4G16360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117731.7 | NP_193369.3 | 1 - 2 | 644 | 65 | |
Set Membership: |
|