Assay Details
Gene Symbol: AT4G16442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117740.5 | NP_567497.1 | 1 - 2 | 463 | 70 | |
GenBank mRNA | AY088534.1 | - | 1 - 2 | 287 | 70 | |
BT003927.1 | - | 1 - 2 | 287 | 70 | ||
BT006082.1 | - | 1 - 2 | 168 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
