Assay Details
Gene Symbol: AT4G16442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117740.5 | NP_567497.1 | 2 - 3 | 590 | 101 | |
GenBank mRNA | AY088534.1 | - | 2 - 3 | 414 | 101 | |
BT003927.1 | - | 2 - 3 | 414 | 101 | ||
BT006082.1 | - | 2 - 3 | 295 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
