Assay Details
| Gene Symbol: AT4G16560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117755.1 | NP_567505.1 | 15 - 16 | 1269 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G16560| Gene Symbol: AT4G16560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117755.1 | NP_567505.1 | 15 - 16 | 1269 | 106 | |
Set Membership: |
|