Assay Details
| Gene Symbol: PSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117771.3 | NP_193403.2 | 10 - 11 | 1250 | 65 | |
| GenBank mRNA | AY189805.1 | - | 10 - 11 | 1072 | 65 | |
| BT026135.1 | - | 10 - 11 | 1072 | 65 | ||
More Information
Set Membership: |
|

PSD1
Genomic Map