Assay Details
| Gene Symbol: PSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341129.1 | NP_001329522.1 | 2 - 3 | 391 | 80 | |
| NM_117771.3 | NP_193403.2 | 2 - 3 | 470 | 80 | ||
| GenBank mRNA | AY189805.1 | - | 2 - 3 | 292 | 80 | |
| BT026135.1 | - | 2 - 3 | 292 | 80 | ||
More Information
Set Membership: |
|

PSD1
Genomic Map