Assay Details
| Gene Symbol: PSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341129.1 | NP_001329522.1 | 6 - 7 | 680 | 69 | |
| NM_117771.3 | NP_193403.2 | 6 - 7 | 759 | 69 | ||
| GenBank mRNA | AY189805.1 | - | 6 - 7 | 581 | 69 | |
| BT026135.1 | - | 6 - 7 | 581 | 69 | ||
More Information
Set Membership: |
|

PSD1
Genomic Map