Assay Details
| Gene Symbol: CT-BMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117813.3 | NP_567523.1 | 3 - 4 | 1001 | 83 | |
| GenBank mRNA | AY087592.1 | - | 3 - 4 | 933 | 83 | |
More Information
Set Membership: |
|

CT-BMY
Genomic Map