Assay Details
Gene Symbol: RABB1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117821.4 | NP_193450.1 | 4 - 5 | 657 | 153 | |
GenBank mRNA | BT002513.1 | - | 4 - 5 | 572 | 153 | |
BT006251.1 | - | 4 - 5 | 469 | 153 | ||
Y09314.1 | - | 583 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
