Assay Details
Gene Symbol: NS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341198.1 | NP_001328216.1 | 7 - 8 | 1179 | 75 | |
NM_117835.5 | NP_193462.1 | 7 - 8 | 1011 | 75 | ||
GenBank mRNA | AY078967.1 | - | 7 - 8 | 988 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
