Assay Details
Gene Symbol: UCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117857.3 | NP_193484.1 | 3 - 4 | 437 | 91 | |
GenBank mRNA | AK118904.1 | - | 3 - 4 | 376 | 91 | |
BT005488.1 | - | 3 - 4 | 254 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
