Assay Details
Gene Symbol: UCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117857.3 | NP_193484.1 | 6 - 7 | 680 | 142 | |
GenBank mRNA | AK118904.1 | - | 6 - 7 | 619 | 142 | |
BT005488.1 | - | 6 - 7 | 497 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
