Assay Details
Gene Symbol: UCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117857.3 | NP_193484.1 | 8 - 9 | 803 | 84 | |
GenBank mRNA | AK118904.1 | - | 8 - 9 | 742 | 84 | |
BT005488.1 | - | 8 - 9 | 620 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
