Assay Details
| Gene Symbol: CBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341236.1 | NP_001329533.1 | 2 - 3 | 464 | 84 | |
| NM_001341237.1 | NP_001329535.1 | 2 - 3 | 534 | 84 | ||
| NM_001341238.1 | NP_001329534.1 | 2 - 3 | 274 | 84 | ||
| NM_117870.4 | NP_567533.1 | 2 - 3 | 540 | 84 | ||
| NM_202837.1 | NP_974566.1 | 2 - 3 | 252 | 84 | ||
| GenBank mRNA | AF076251.1 | - | 2 - 3 | 270 | 84 | |
| AK118798.1 | - | 2 - 3 | 345 | 84 | ||
| BT005567.1 | - | 1 - 2 | 84 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_202837.1 transcripts.
Set Membership: |
|

CBL1
Genomic Map