Assay Details
| Gene Symbol: CBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341236.1 | NP_001329533.1 | 4 - 5 | 592 | 109 | |
| NM_001341237.1 | NP_001329535.1 | 4 - 5 | 662 | 109 | ||
| NM_001341238.1 | NP_001329534.1 | 4 - 5 | 402 | 109 | ||
| NM_117870.4 | NP_567533.1 | 4 - 5 | 668 | 109 | ||
| NM_202837.1 | NP_974566.1 | 3 - 4 | 679 | 109 | ||
| GenBank mRNA | AF076251.1 | - | 4 - 5 | 398 | 109 | |
| AK118798.1 | - | 4 - 5 | 473 | 109 | ||
| BT005567.1 | - | 3 - 4 | 212 | 109 | ||
More Information
Set Membership: |
|

CBL1
Genomic Map