Assay Details
| Gene Symbol: CBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341236.1 | NP_001329533.1 | 5 - 6 | 714 | 69 | |
| NM_001341237.1 | NP_001329535.1 | 5 - 6 | 784 | 69 | ||
| NM_001341238.1 | NP_001329534.1 | 5 - 6 | 524 | 69 | ||
| NM_117870.4 | NP_567533.1 | 5 - 6 | 790 | 69 | ||
| NM_202837.1 | NP_974566.1 | 4 - 5 | 801 | 69 | ||
| GenBank mRNA | AF076251.1 | - | 5 - 6 | 520 | 69 | |
| AK118798.1 | - | 5 - 6 | 595 | 69 | ||
| BT005567.1 | - | 4 - 5 | 334 | 69 | ||
More Information
Set Membership: |
|

CBL1
Genomic Map