Assay Details
| Gene Symbol: CBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341236.1 | NP_001329533.1 | 6 - 7 | 769 | 87 | |
| NM_001341237.1 | NP_001329535.1 | 6 - 7 | 839 | 87 | ||
| NM_001341238.1 | NP_001329534.1 | 6 - 7 | 579 | 87 | ||
| NM_117870.4 | NP_567533.1 | 6 - 7 | 845 | 87 | ||
| NM_202837.1 | NP_974566.1 | 5 - 6 | 856 | 87 | ||
| GenBank mRNA | AF076251.1 | - | 6 - 7 | 575 | 87 | |
| AK118798.1 | - | 6 - 7 | 650 | 87 | ||
| BT005567.1 | - | 5 - 6 | 389 | 87 | ||
More Information
Set Membership: |
|

CBL1
Genomic Map