Assay Details
Gene Symbol: SYP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084934.1 | NP_001078403.1 | 3 - 4 | 685 | 93 | |
NM_117882.3 | NP_567537.1 | 3 - 4 | 685 | 93 | ||
GenBank mRNA | AK316791.1 | - | 3 - 4 | 643 | 93 | |
AY074294.1 | - | 3 - 4 | 643 | 93 | ||
BT000708.1 | - | 3 - 4 | 642 | 93 | ||
BT008517.1 | - | 3 - 4 | 437 | 93 | ||
U85036.1 | - | 3 - 4 | 612 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
