Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117914.4 | NP_193538.1 | 1 - 2 | 377 | 65 | |
| GenBank mRNA | AB107250.1 | - | 1 - 2 | 329 | 65 | |
| AF389286.1 | - | 1 - 2 | 328 | 65 | ||
| AY086496.1 | - | 1 - 2 | 331 | 65 | ||
| AY093750.1 | - | 1 - 2 | 296 | 65 | ||
| Y10548.1 | - | 323 | 65 | |||
More Information
Set Membership: |
|

EIF4E
Genomic Map