Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117914.4 | NP_193538.1 | 4 - 5 | 729 | 101 | |
| GenBank mRNA | AB107250.1 | - | 4 - 5 | 681 | 101 | |
| AF389286.1 | - | 4 - 5 | 680 | 101 | ||
| AY086496.1 | - | 4 - 5 | 683 | 101 | ||
| AY093750.1 | - | 4 - 5 | 648 | 101 | ||
| Y10548.1 | - | 675 | 101 | |||
More Information
Set Membership: |
|

EIF4E
Genomic Map