Assay Details
| Gene Symbol: SSP4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036584.3 | NP_001031661.2 | 6 - 7 | 1427 | 89 | |
| NM_117924.7 | NP_193548.7 | 5 - 6 | 1650 | 89 | ||
More Information
Set Membership: |
|

SSP4b
Genomic Map