Assay Details
Gene Symbol: SS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117934.4 | NP_193558.3 | 14 - 15 | 2979 | 110 | |
GenBank mRNA | AK226610.1 | - | 14 - 15 | 2984 | 110 | |
KJ676990.1 | - | 14 - 15 | 2866 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
