Assay Details
Gene Symbol: SS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117934.4 | NP_193558.3 | 8 - 9 | 2169 | 74 | |
GenBank mRNA | AK226610.1 | - | 8 - 9 | 2174 | 74 | |
KJ676990.1 | - | 8 - 9 | 2056 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
