Assay Details
Gene Symbol: CHLI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117962.3 | NP_193583.1 | 1 - 2 | 209 | 79 | |
GenBank mRNA | AK227146.1 | - | 1 - 2 | 116 | 79 | |
AY093192.1 | - | 1 - 2 | 152 | 79 | ||
BT010129.1 | - | 1 - 2 | 101 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
