Assay Details
Gene Symbol: PNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203432.1 | NP_001190361.1 | 4 - 5 | 1321 | 69 | |
NM_001343720.1 | NP_001332623.1 | 3 - 4 | 1022 | 69 | ||
NM_147894.4 | NP_680199.1 | 4 - 5 | 1261 | 69 | ||
GenBank mRNA | AK226614.1 | - | 4 - 5 | 1266 | 69 | |
AY302133.1 | - | 3 - 4 | 1014 | 69 | ||
BT022100.1 | - | 4 - 5 | 1166 | 69 | ||
BT026521.1 | - | 4 - 5 | 1040 | 69 | ||
KJ138673.1 | - | 4 - 5 | 1040 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
