Assay Details
| Gene Symbol: FAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198391.1 | NP_001185320.1 | 3 - 4 | 891 | 98 | |
| NM_105221.3 | NP_176725.1 | 3 - 4 | 918 | 98 | ||
| GenBank mRNA | AY126994.1 | - | 3 - 4 | 867 | 98 | |
More Information
Set Membership: |
|

FAS1
Genomic Map